Invitrogen
Manufacturer Code:PA537121
Catalog # PIPA537121
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to residues in human AGRN |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Agrin; Agrin Agrin N-terminal 110 kDa subunit Agrin C-terminal 110 kDa subunit Agrin C-terminal 90 kDa fragment C90 Agrin C-terminal 22 kDa fragment C22 AGRN AGRIN; Agrin C-terminal 110 kDa subunit; Agrin C-terminal 22 kDa fragment; Agrin C-terminal 90 kDa fragment; Agrin N-terminal 110 kDa subunit; agrin proteoglycan; C22; C90
Gene Aliases: AGR; AGRIN; AGRN; CMS8; CMSPPD; nmf380
UniProt ID: (Human) O00468, (Rat) P25304
Entrez Gene ID: (Human) 375790, (Mouse) 11603, (Rat) 25592
Molecular Function:
extracellular matrix linker protein
extracellular matrix protein
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.