Novus Biologicals
Manufacturer Code:NBP290675100UL
Catalog # NB025097
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 36 |
Immunogen | Recombinant Mouse Ago2/eIF2C2 protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: AGO2argonaute 2 argonaute2 EC 3.1.26.n1 EC 3.1.26.n2 eIF2C 2 eIF-2C 2 Eukaryotic translation initiation factor 2C 2 eukaryotic translation initiation factor 2C 2 hAgo2 MGC3183 PAZ Piwi domain protein PPD protein argonaute-2 Protein slicer Q10; argonaute 2; Argonaute RISC catalytic component 2; Argonaute2; CTA-204B4.6; eIF-2C 2; eIF2C 2; Eukaryotic translation initiation factor 2C 2; eukaryotic translation initiation factor 2C, 2; hAgo2; PAZ Piwi domain protein; PPD; Protein argonaute-2; Protein slicer
Gene Aliases: AGO2; EIF2C2; Q10
UniProt ID: (Human) Q9UKV8
Entrez Gene ID: (Human) 27161
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.