Novus Biologicals
Manufacturer Code:NB12011570
Catalog # NB12011570
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | CS-56 |
Immunogen | Ventral membranes of chicken gizzard fibroblasts. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: AGC1SEDK aggrecan aggrecan core protein Cartilage-specific proteoglycan core protein Chondroitin sulfate proteoglycan 1 Chondroitin sulfate proteoglycan core protein 1 CSPCP CSPG1aggrecan 1 CSPGCP large aggregating proteoglycan MSK16AGCAN; Aggrecan core protein; Aggrecan core protein 2; Cartilage-specific proteoglycan core protein; Chondroitin sulfate proteoglycan 1; Chondroitin sulfate proteoglycan core protein 1; CSPCP; large aggregating proteoglycan
Gene Aliases: ACAN; AGC1; AGCAN; CSPG1; CSPGCP; MSK16; SEDK
UniProt ID: (Human) P16112
Entrez Gene ID: (Human) 176
Molecular Function:
extracellular matrix glycoprotein
extracellular matrix protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.