Invitrogen
Manufacturer Code:MA316888
Catalog # AMSA6888
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | BC-3 |
Immunogen | Synthetic peptide corresponding to human aggrecan ARGSV neoepitope. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: aggrecan 1; aggrecan 1 (chondroitin sulfate proteoglycan 1, large aggregating proteoglycan, antigen identified by monoclonal antibody A0122); aggrecan AGC1 CSPG1 MSK16 CSPGCP Chondroitin sulfate proteoglycan 1 Cartilage-specific proteoglycan core protein Chondroitin sulfate proteoglycan core protein 1 SEDK aggrecan 1 AGCAN aggrecan core protein large aggregating proteoglycan CSPCP; Aggrecan core protein; Aggrecan core protein 2; aggrecan epidermal growth factor-like domain-1; aggrecan interglobular domain; aggrecan, structural proteoglycan of cartilage; cartilage aggregating proteoglycan; Cartilage-specific proteoglycan core protein; Chondroitin sulfate proteoglycan 1; Chondroitin sulfate proteoglycan core protein 1; CSPCP; high density cartilage proteoglycan; large aggregating proteoglycan; large aggregating proteoglycan, antigen
Gene Aliases: ACAN; Agc; AGC1; AGCAN; CSPCP; CSPG1; CSPGCP; MSK16; SEDK
UniProt ID: (Bovine) P13608, (Human) P16112, (Rat) P07897
Entrez Gene ID: (Dog) 403828, (Rabbit) 100009079, (Pig) 397255, (Horse) 100033876, (Bovine) 280985, (Human) 176, (Sheep) 443001, (Guinea pig) 100731672, (Rat) 58968
Molecular Function:
extracellular matrix glycoprotein
extracellular matrix protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.