Novus Biologicals
Manufacturer Code:NBP255539
Catalog # NBP255539
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:LPLPRNITEGEARGSVILTVKPIFEVSPSPLEPEEPFTFAPEIGATAFAEVENETGEATRPWGFPTPG |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AGC1SEDK aggrecan aggrecan core protein Cartilage-specific proteoglycan core protein Chondroitin sulfate proteoglycan 1 Chondroitin sulfate proteoglycan core protein 1 CSPCP CSPG1aggrecan 1 CSPGCP large aggregating proteoglycan MSK16AGCAN; Aggrecan core protein; Aggrecan core protein 2; Cartilage-specific proteoglycan core protein; Chondroitin sulfate proteoglycan 1; Chondroitin sulfate proteoglycan core protein 1; CSPCP; large aggregating proteoglycan
Gene Aliases: ACAN; AGC1; AGCAN; CSPG1; CSPGCP; MSK16; SEDK
UniProt ID: (Human) P16112
Entrez Gene ID: (Human) 176
Molecular Function:
extracellular matrix glycoprotein
extracellular matrix protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.