R&D Systems
Manufacturer Code:AF1220
Catalog # AF1220
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Mouse myeloma cell line NS0-derived recombinant human Aggrecan Val20-Arg675 Accession # NP_037359 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: ACAN AGC1 AGC1SEDK aggrecan aggrecan core protein Cartilage-specific proteoglycan core protein Chondroitin sulfate proteoglycan 1 Chondroitin sulfate proteoglycan core protein 1 CSPCP CSPG1 CSPG1aggrecan 1 CSPGCP large aggregating proteoglycan MSK16 MSK16AGCAN SEDK; Aggrecan core protein; Aggrecan core protein 2; Cartilage-specific proteoglycan core protein; Chondroitin sulfate proteoglycan 1; Chondroitin sulfate proteoglycan core protein 1; CSPCP; large aggregating proteoglycan
Gene Aliases: ACAN; AGC1; AGCAN; CSPG1; CSPGCP; MSK16; SEDK
UniProt ID: (Human) P16112
Entrez Gene ID: (Human) 176
Molecular Function:
extracellular matrix glycoprotein
extracellular matrix protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.