Proteintech
Manufacturer Code:138801AP150UL
Catalog # 50-172-9061
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Aggrecan Fusion Protein Ag4985 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ACAN AGC1 AGCAN aggrecan CSPG1 CSPGCP MSK16 SEDK; aggrecan 1; Aggrecan core protein; Aggrecan core protein 2; aggrecan, structural proteoglycan of cartilage; Cartilage-specific proteoglycan core protein; Chondroitin sulfate proteoglycan 1; Chondroitin sulfate proteoglycan core protein 1; CSPCP; large aggregating proteoglycan
Gene Aliases: ACAN; Agc; AGC1; AGCAN; b2b183Clo; cmd; CSPG1; CSPGCP; MSK16; SEDK
UniProt ID: (Human) P16112, (Rat) P07897, (Mouse) Q61282
Entrez Gene ID: (Human) 176, (Rat) 58968, (Mouse) 11595
Molecular Function: extracellular matrix glycoprotein extracellular matrix protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.