Novus Biologicals
Manufacturer Code:NBP290199AF532
Catalog # NB070873
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 120 |
Immunogen | This antibody was obtained from a rabbit immunized with purified recombinant Human Afamin (P43652 Met1-Asn599). |
Conjugate | Alexa Fluor 532 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: Afamin; afamin ALB2alpha-Alb ALBAalpha-albumin ALF Alpha-Alb Alpha-albumin MGC125338 MGC125339; Alpha-Alb; Alpha-albumin
Gene Aliases: AFM; ALB2; ALBA; ALF
UniProt ID: (Human) Q32MR3
Entrez Gene ID: (Human) 173
Molecular Function:
transfer/carrier protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.