R&D Systems
Manufacturer Code:AF8119SP
Catalog # AF8119SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Sheep |
Class | Polyclonal |
Type | Antibody |
Immunogen | Mouse myeloma cell line NS0-derived recombinant mouse Afamin Leu22-Met608 Accession # O89020 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Sheep Polyclonal Antibody
For Research Use Only
Protein Aliases: Afamin; afamin AFM ALB2 ALB2alpha-Alb ALBA ALBAalpha-albumin ALF Alpha-Alb Alpha-albumin MGC125338 MGC125339; Alpha-Alb; Alpha-albumin
Gene Aliases: AFM; ALB2; ALBA; ALF
UniProt ID: (Human) Q32MR3
Entrez Gene ID: (Human) 173
Molecular Function:
transfer/carrier protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.