R&D Systems
Manufacturer Code:FAB10293V100UG
Catalog # FAB10293V10
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 528563 |
Immunogen | HEK293 human embryonic kidney cell line transfected with human Adrenomedullin R/ADMR Accession &num O15218 |
Conjugate | Alexa Fluor 405 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Protect from light. Do not freeze. 12 months from date of receipt 2 to 8°C as supplied. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 7TMR ADMR adrenomedullin receptor AdrenomedullinR AMR AM-R G protein-coupled receptor 1827TMR G10D GAMRH Gpcr22 GPR182 G-protein coupled receptor 182 hrhAMR MB10 MGC34399 NOW; adrenomedullin receptor; G-protein coupled receptor 182
Gene Aliases: 7TMR; ADMR; AM-R; AMR; G10D; gamrh; GPR182; hrhAMR
UniProt ID: (Human) O15218
Entrez Gene ID: (Human) 11318
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.