Invitrogen
Manufacturer Code:PA536524
Catalog # PIPA536524
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to amino acids 70-120 of human ADM |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ADM; Adrenomedullin; Adrenomedullin Proadrenomedullin N-20 terminal peptide ProAM N-terminal 20 peptide PAMP ProAM-N20 ADM AM; AM; PAMP; preproadrenomedullin; Pro-adrenomedullin; Proadrenomedullin N-20 terminal peptide; ProAM N-terminal 20 peptide
Gene Aliases: ADM; AM; Ap; H39316; PAMP; RATAP
UniProt ID: (Human) P35318, (Rat) P43145, (Mouse) P97297
Entrez Gene ID: (Human) 133, (Rat) 25026, (Mouse) 11535
Molecular Function: peptide hormone signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.