R&D Systems
Manufacturer Code:MAB61081100
Catalog # MAB61081100
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1003019 |
Immunogen | Human Adrenomedullin/ADM synthetic peptide Accession # P35318 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70°C as supplied. 1 month 2 to 8°C under sterile conditions after reconstitution. 6 months -20 to -70°C under sterile conditions a |
Mouse Monoclonal Antibody
Protein Aliases: ADM adrenomedullin AMpreproadrenomedullin; Adrenomedullin; AM; PAMP; preproadrenomedullin; Pro-adrenomedullin; Proadrenomedullin N-20 terminal peptide; ProAM N-terminal 20 peptide
Gene Aliases: ADM; AM; PAMP
UniProt ID: (Human) P35318
Entrez Gene ID: (Human) 133
Molecular Function: peptide hormone signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.