Invitrogen
Manufacturer Code:PA560038
Catalog # PIPA560038
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human AdoHcyase 2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: adenosylhomocysteinase-like 1; AdoHcyase 2; DC-expressed AHCY-like molecule; DCAL IRBIT PRO0233 XPVKONA DC-expressed AHCY-like molecule S-adenosyl homocysteine hydrolase homolog S-adenosyl-L-homocysteine hydrolase 2 S-adenosylhomocysteine hydrolase-like protein 1 adoHcyase 2 dendritic cell expressed AHCY-like protein inositol 145-trisphosphate receptor-bi; dendritic cell expressed AHCY-like protein; inositol 1,4,5-trisphosphate receptor-binding protein; IP(3)Rs binding protein released with IP(3); IP3R binding protein released with inositol 1,4,5-trisphosphate; IP3R-binding protein released with inositol 1,4,5-trisphosphate; IRBIT; protein phosphatase 1, regulatory subunit 78; Putative adenosylhomocysteinase 2; S-adenosyl homocysteine hydrolase homolog; S-adenosyl-L-homocysteine hydrolase 2; S-adenosylhomocysteine hydrolase, related sequence 3; S-adenosylhomocysteine hydrolase-like 1; S-adenosylhomocysteine hydrolase-like protein 1
Gene Aliases: 1110034F20Rik; AA409031; AA414901; Ahcy-rs3; AHCYL1; DCAL; IRBIT; PPP1R78; PRO0233; XPVKONA
UniProt ID: (Human) O43865, (Rat) B5DFN2, (Mouse) Q80SW1
Entrez Gene ID: (Human) 10768, (Rat) 362013, (Mouse) 229709
Molecular Function: hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.