Invitrogen
Manufacturer Code:PA5112298
Catalog # PIPA5112298
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | E. coli-derived Human AHCYL1/IRBIT fragment |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 1110034F20Rik AA409031 AA414901 adenosylhomocysteinase 2 adenosylhomocysteinase like 1 adenosylhomocysteinase-like 1 AdoHcyase 2 Ahcyl1 Ahcyl1-v1 Ahcyl1-v2 Ahcy-rs3 BOS_3128 cb586 DCAL DC-expressed AHCY-like molecule dendritic cell expressed AHCY-like protein inositol 145-trisphosphate receptor-binding protein IP(3)Rs binding protein released with IP(3) IP3R binding protein released with inositol 145-trisphosphate IP3R-binding protein released with inositol 145-trisphosphate Irbit PPP1R78 PRO0233 protein phosphatase 1 regulatory subunit 78 Putative adenosylhomocysteinase 2 s adenosylhomocysteine hydrolase S-adenosyl homocysteine hydrolase homolog S-adenosylhomocysteine hydrolase related sequence 3 S-adenosylhomocysteine hydrolase-like 1 S-adenosylhomocysteine hydrolase-like protein 1 S-adenosylhomocysteine hydrolase-like protein variant 1 S-adenosylhomocysteine hydrolase-like protein variant 2 S-adenosyl-L-homocysteine hydrolase 2 wu:fj66f02 XPVKONA; adenosylhomocysteinase-like 1; AdoHcyase 2; DC-expressed AHCY-like molecule; dendritic cell expressed AHCY-like protein; inositol 1,4,5-trisphosphate receptor-binding protein; IP(3)Rs binding protein released with IP(3); IRBIT; protein phosphatase 1, regulatory subunit 78; Putative adenosylhomocysteinase 2; S-adenosyl homocysteine hydrolase homolog; S-adenosyl-L-homocysteine hydrolase 2; S-adenosylhomocysteine hydrolase-like protein 1
Gene Aliases: AHCYL1; DCAL; IRBIT; PPP1R78; PRO0233; XPVKONA
UniProt ID: (Human) O43865
Entrez Gene ID: (Human) 10768
Molecular Function:
hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.