R&D Systems
Manufacturer Code:AF5208SP
Catalog # AF5208SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Sheep |
Class | Polyclonal |
Type | Antibody |
Immunogen | <EM>E. coli</EM>-derived recombinant human Adiponutrin/PNPLA3 Arg160-Arg349 Accession # Q9NST1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Sheep Polyclonal Antibody
For Research Use Only
Protein Aliases: 1-acylglycerol-3-phosphate O-acyltransferase PNPLA3; acylglycerol O-acyltransferase; Acylglycerol O-acyltransferase adiponutrin ADPN ADPNdJ796I17.1 C22orf20 Calcium-independent phospholipase A2-epsilon chromosome 22 open reading frame 20 EC 2.3.1.- EC 2.7.7.56 EC 3.1.1.3 EC 4.2.3.4 FLJ22012 iPLA(2)epsilon iPLA2-epsilon patatin-like phospholipase domain containing 3 patatin-like phospholipase domain-containing protein 3 PNPLA3; Acylglycerol transacylase; Adiponutrin; ADPN; Calcium-independent phospholipase A2-epsilon; iPLA2-epsilon; iPLA2epsilon; Lysophosphatidic acid acyltransferase; patatin-like phospholipase domain containing 3; Patatin-like phospholipase domain-containing protein 3
Gene Aliases: ADPN; C22orf20; iPLA(2)epsilon; PNPLA3
UniProt ID: (Human) Q9NST1
Entrez Gene ID: (Human) 80339
Molecular Function:
acyltransferase
hydrolase
lipase
phospholipase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.