Proteintech
Manufacturer Code:143611AP150UL
Catalog # 50-172-9397
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Adiponectin receptor Fusion Protein Ag5744 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ACDCR2 adiponectin receptor 2 Adiponectin receptor protein 2 ADIPOR1 ADIPOR2 PAQR2; Adiponectin receptor protein 1; Adiponectin receptor protein 2; Progestin and adipoQ receptor family member 1; Progestin and adipoQ receptor family member 2; Progestin and adipoQ receptor family member I; Progestin and adipoQ receptor family member II
Gene Aliases: 1110001I14Rik; 2810031L11Rik; ACDCR1; ACDCR2; ADCR2; ADIPOR1; ADIPOR2; AI115388; AW554121; CGI-45; CGI45; D6Ucla1e; PAQR1; PAQR2; Parq1; Parq2; TESBP1A
UniProt ID: (Human) Q96A54, (Human) Q86V24, (Mouse) Q91VH1, (Mouse) Q8CA45
Entrez Gene ID: (Human) 51094, (Human) 79602, (Rat) 289036, (Mouse) 72674, (Rat) 312670, (Mouse) 68465
Molecular Function: receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.