Invitrogen
Manufacturer Code:PA560488
Catalog # PIPA560488
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human Adiponectin Receptor 2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Adiponectin receptor protein 2; ADIPOR2 ACDCR2 Adiponectin receptor protein 2 FLJ21432 MGC4640 PAQR2 Progestin and adipoQ receptor family member II; Progestin and adipoQ receptor family member 2; Progestin and adipoQ receptor family member II
Gene Aliases: ACDCR2; ADIPOR2; PAQR2
UniProt ID: (Human) Q86V24
Entrez Gene ID: (Human) 79602
Molecular Function:
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.