Invitrogen
Manufacturer Code:PA578288
Catalog # PIPA578288
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Full length human Adiponectin Recombit protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 30 kDa adipocyte complement-related protein; ACDC ACRP30 ADIPQTL1 ADPN APM-1 APM1 GBP28 adiponectin ADIPOQ adipocyte complement-related 30 kDa protein 30 kDa adipocyte complement-related protein gelatin-binding protein 28 adipose specific collagen-like factor adipose most abundant gene transcript 1 protein "adiponectin C1Q and collagen domain containing " ACRP30; adipocyte complement related protein; adipocyte complement related protein of 30 kDa; Adipocyte complement-related 30 kDa protein; adipocyte, C1Q and collagen domain containing; Adipocyte, C1q and collagen domain-containing protein; Adipocyte-specific protein AdipoQ; Adiponectin; Adipose most abundant gene transcript 1 protein; adipose specific collagen-like factor; apM-1; Gelatin-binding protein; gelatin-binding protein 28
Gene Aliases: 30kDa; ACDC; ACRP30; Ad; adipo; ADIPOQ; ADIPQTL1; ADPN; APM-1; APM1; APN; GBP28
UniProt ID: (Human) Q15848, (Mouse) Q60994
Entrez Gene ID: (Human) 9370, (Rat) 246253, (Mouse) 11450
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.