R&D Systems
Manufacturer Code:FAB10128N100UG
Catalog # FB10128N100
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 2416C |
Immunogen | Synthetic peptide containing human AdipoR1 C-term sequences |
Conjugate | Alexa Fluor 700 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Protect from light. Do not freeze. 12 months from date of receipt 2 to 8°C as supplied. |
Rabbit Monoclonal Antibody
Protein Aliases: ACDCR1 adiponectin receptor 1 adiponectin receptor protein 1 ADR1 CGI45 FLJ42464 PAQR1 PAQR1FLJ25385 Progestin and adipoQ receptor family member I TESBP1A; Adiponectin receptor protein 1; Progestin and adipoQ receptor family member 1; Progestin and adipoQ receptor family member I
Gene Aliases: ACDCR1; ADIPOR1; CGI-45; CGI45; PAQR1; TESBP1A
UniProt ID: (Human) Q96A54
Entrez Gene ID: (Human) 51094
Molecular Function:
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.