Proteintech
Manufacturer Code:152641AP150UL
Catalog # 50-173-0102
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Adenylosuccinate lyase Fusion Protein Ag7332 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Adenylosuccinase; Adenylosuccinase adenylosuccinate lyase ADSL AMPS ASASE ASL; Adenylosuccinate lyase; adenylosuccinate lyase 1; ADSL; ASase; ASL; test
Gene Aliases: Adl; ADSL; AMPS; ASASE; ASL; null
UniProt ID: (Human) P30566, (Mouse) P54822
Entrez Gene ID: (Human) 158, (Rat) 315150, (Mouse) 11564
Molecular Function:
lyase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.