Novus Biologicals
Manufacturer Code:NBP29266120UL
Catalog # NB027192
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-280 of human ADSS (NP_001117.2). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ADEH adenylosuccinate synthase adenylosuccinate synthetase (Ade(-)H-complementing) adenylosuccinate synthetase isozyme 2 Adenylosuccinate synthetase acidic isozyme Adenylosuccinate synthetase liver isozyme AdSS 2 ADSS2 AMPSase 2 EC 6.3.4.4 IMP--aspartate ligase 2 L-type adenylosuccinate synthetase MGC20404; adenylosuccinate synthetase (Ade(-)H-complementing); Adenylosuccinate synthetase isozyme 2; Adenylosuccinate synthetase, acidic isozyme; Adenylosuccinate synthetase, liver isozyme; adSS 2; AMPSase 2; IMP--aspartate ligase 2; L-type adenylosuccinate synthetase
Gene Aliases: ADEH; ADSS; ADSS 2; ADSS2
UniProt ID: (Human) P30520
Entrez Gene ID: (Human) 159
Molecular Function: ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.