Novus Biologicals
Manufacturer Code:NBP299767AF532
Catalog # NB062790
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with purified recombinant Adenylosuccinate Lyase (P30566-1 Met1-Leu484). |
Conjugate | Alexa Fluor 532 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Adenylosuccinase; adenylosuccinase adenylosuccinate lyase AMPS ASase ASL EC 4.3.2 EC 4.3.2.2; Adenylosuccinate lyase; ADSL; ASase
Gene Aliases: ADSL; AMPS; ASASE; ASL
UniProt ID: (Human) P30566
Entrez Gene ID: (Human) 158
Molecular Function:
lyase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.