Invitrogen
Manufacturer Code:PA528354
Catalog # PIPA528354
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 1 and 188 of Human Adenylate kinase 1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Adenylate kinase isoenzyme 1; Adenylate monophosphate kinase; AK 1; AK 1 ATP-AMP transphosphorylase 1 adenylate kinase isoenzyme 1 myokinase; ATP-AMP transphosphorylase 1; ATP:AMP phosphotransferase; cytosolic adenylate kinase; Myokinase; testis secretory sperm binding protein Li 58j
Gene Aliases: Ak-1; AK1; B430205N08Rik; HTL-S-58j
UniProt ID: (Human) P00568, (Mouse) Q9R0Y5
Entrez Gene ID: (Human) 203, (Mouse) 11636
Molecular Function: kinase nucleotide kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.