Invitrogen
Manufacturer Code:PA589416
Catalog # PIPA589416
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fusion protein containing a sequence corresponding to amino acids 1-200 of human AK8 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Adenylate kinase 8; Adenylate kinase isoenzyme 4, mitochondrial (ATP-AMP transphosphorylase); AK 8; ATP-AMP transphosphorylase 8; HypB, AroK, ADK and rho factor domain containing protein RGD1303144; putative adenylate kinase-like protein C9orf98 homolog
Gene Aliases: 1190002A17Rik; AK 8; Ak8; RGD1303144
UniProt ID: (Rat) Q68FP8, (Mouse) Q32M07
Entrez Gene ID: (Rat) 311833, (Mouse) 68870
Molecular Function:
kinase
nucleotide kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.