Invitrogen
Manufacturer Code:PA554918
Catalog # PIPA554918
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human Adenylate Kinase 8 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Adenylate kinase 8; AK 8; ATP-AMP transphosphorylase 8; C9orf98 DDX31 RP11-143F18.1 putative adenylate kinase-like protein C9orf98; putative adenylate kinase-like protein C9orf98
Gene Aliases: AK 8; AK8; C9orf98; DDX31
UniProt ID: (Human) Q96MA6
Entrez Gene ID: (Human) 158067
Molecular Function: kinase nucleotide kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.