Invitrogen
Manufacturer Code:PA515429
Catalog # PIPA515429
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 1-30 amino acids from the N-terminal region of human AK2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Adenylate kinase 2, mitochondrial; Adenylate kinase 2, mitochondrial, N-terminally processed; Adenylate kinase isoenzyme 2 ATP-AMP transphosphorylase; adenylate kinase isoenzyme 2, mitochondrial; Adenylate monophosphate kinase; AK 2; ATP-AMP transphosphorylase 2; ATP:AMP phosphotransferase; testis secretory sperm-binding protein Li 220n
Gene Aliases: ADK2; Ak-2; AK2; D4Ertd220e
UniProt ID: (Human) P54819, (Mouse) Q9WTP6
Entrez Gene ID: (Human) 204, (Mouse) 11637
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.