Invitrogen
Manufacturer Code:PA5116423
Catalog # PIPA5116423
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Zebrafish |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 2-36 amino acids from the N-terminal region of (DANRE) ak2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: adenylate kinase 2 adenylate kinase 2 mitochondrial adenylate kinase 2 mitochondrial {ECO:0000255 Adenylate kinase 2 mitochondrial N-terminally processed adenylate kinase isoenzyme 2 mitochondrial Adenylate monophosphate kinase adenylate monophosphate kinase {ECO:0000255 ADK2 AK 2 AK 2 {ECO:0000255 AK2 Ak-2 ATP:AMP phosphotransferase ATP:AMP phosphotransferase {ECO:0000255 ATP-AMP transphosphorylase 2 ATP-AMP transphosphorylase 2 {ECO:0000255 D4Ertd220e HAMAP-Rule:MF_03168} si:ch211-197n10.1 testis secretory sperm-binding protein Li 220n wu:fb34f05 wu:fj80e03; Adenylate kinase 2, mitochondrial; Adenylate monophosphate kinase; AK 2; ATP-AMP transphosphorylase 2; ATP:AMP phosphotransferase
Gene Aliases: ak2; si:ch211-197n10.1; wu:fb34f05; wu:fj80e03
UniProt ID: (Zebrafish) Q1L8L9
Entrez Gene ID: (Zebrafish) 321793
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.