Novus Biologicals
Manufacturer Code:NBP303019
Catalog # NB092896
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human Adenylate Cyclase 8. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. Do not freeze. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AC8; AC8 ADCY3 adenylate cyclase 8 (brain) adenylate cyclase type 8 Adenylate cyclase type VIII Adenylyl cyclase 8 adenylyl cyclase-8 brain ATP pyrophosphate-lyase 8 Ca(2+)/calmodulin-activated adenylyl cyclase EC 4.6.1.1 HBAC1; adenylate cyclase 8 (brain); Adenylate cyclase type 8; Adenylate cyclase type VIII; Adenylyl cyclase 8; adenylyl cyclase-8, brain; ATP pyrophosphate-lyase 8; Ca(2+)/calmodulin-activated adenylyl cyclase; epididymis secretory sperm binding protein Li 172mP; HEL-S-172mP
Gene Aliases: AC8; ADCY3; ADCY8; HBAC1
UniProt ID: (Human) P40145
Entrez Gene ID: (Human) 114
Molecular Function: adenylate cyclase cyclase guanylate cyclase lyase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.