Novus Biologicals
Manufacturer Code:NBP297909R
Catalog # NB049339
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with E. coli-derived Human Adenylate Cyclase 7 fragment. |
Conjugate | DyLight 550 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AC7 adenylate cyclase 7 adenylate cyclase type 7 Adenylate cyclase type VII Adenylyl cyclase 7 ATP pyrophosphate-lyase 7 EC 4.6.1 EC 4.6.1.1 KIAA0037FLJ36387; Adenylate cyclase type 7; Adenylate cyclase type VII; Adenylyl cyclase 7; ATP pyrophosphate-lyase 7
Gene Aliases: AC7; ADCY7; KIAA0037
UniProt ID: (Human) P51828
Entrez Gene ID: (Human) 113
Molecular Function: adenylate cyclase cyclase guanylate cyclase lyase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.