Invitrogen
Manufacturer Code:PA575274
Catalog # PIPA575274
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to amino acids 1111-1160 of Human ADCY 5 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: A cyclase V A cyclase VI Adenylate cyclase type 5 ATP pyrophosphate-lyase 5 Adenylate cyclase type V Adenylyl cyclase 5 ADCY5 Adenylate cyclase type 6 ATP pyrophosphate-lyase 6 Adenylate cyclase type VI Adenylyl cyclase 6 Ca(2+)-inhibitable adenylyl cyclase ADCY6 KIAA0422; AC5; AC6; ACVI; adcy5; Adenylate cyclase type 5; Adenylate cyclase type 6; Adenylate cyclase type V; Adenylate cyclase type VI; Adenylyl cyclase 5; Adenylyl cyclase 6; adenylyl cyclase type V; ATP pyrophosphate-lyase 5; ATP pyrophosphate-lyase 6; Ca(2+)-inhibitable adenylyl cyclase
Gene Aliases: AC5; AC6; ACVI; ADCY5; ADCY6; ADCYB; AW121902; FDFM; KIAA0422; LCCS8; mKIAA0422
UniProt ID: (Human) O95622, (Human) O43306, (Rat) Q04400, (Mouse) P84309, (Rat) Q03343, (Mouse) Q01341
Entrez Gene ID: (Human) 111, (Human) 112, (Rat) 64532, (Mouse) 224129, (Rat) 25289, (Mouse) 11512
Molecular Function: adenylate cyclase cyclase guanylate cyclase lyase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.