Invitrogen
Manufacturer Code:PA538844
Catalog # PIPA538844
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide derived from the internal region of human ADCY5/6 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AC5; AC6; ACVI; adcy5; Adenylate cyclase type 5; Adenylate cyclase type 6; Adenylate cyclase type V; Adenylate cyclase type VI; Adenylyl cyclase 5; Adenylyl cyclase 6; adenylyl cyclase type V; ATP pyrophosphate-lyase 5; ATP pyrophosphate-lyase 6; Ca(2+)-inhibitable adenylyl cyclase
Gene Aliases: AC5; AC6; ACVI; ADCY5; ADCY6; ADCYB; AW121902; FDFM; KIAA0422; LCCS8; mKIAA0422
UniProt ID: (Human) O95622, (Human) O43306, (Rat) Q04400, (Mouse) P84309, (Rat) Q03343, (Mouse) Q01341
Entrez Gene ID: (Human) 111, (Human) 112, (Rat) 64532, (Mouse) 224129, (Rat) 25289, (Mouse) 11512
Molecular Function:
adenylate cyclase
cyclase
guanylate cyclase
lyase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.