Invitrogen
Manufacturer Code:PA572985
Catalog # PIPA572985
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A 20 amino acid peptide corresponding to the C-terminus of rat Adenylate Cyclase 3. &lsqbUniProtNo. P21932] |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AC-III; AC3; AC3 KIAA0511; Adenylate cyclase type 3; Adenylate cyclase type III; Adenylate cyclase, olfactive type; Adenylyl cyclase 3; adenylyl cyclase, type III; ATP pyrophosphate-lyase 3; type III adenylyl cyclase
Gene Aliases: AC-III; AC3; ADCY3; KIAA0511
UniProt ID: (Human) O60266, (Rat) P21932
Entrez Gene ID: (Human) 109, (Rat) 64508
Molecular Function: adenylate cyclase cyclase guanylate cyclase lyase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.