Novus Biologicals
Manufacturer Code:NBP192683F
Catalog # NBP192683F
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | C-terminal peptide of rat ACIII PAAFPNGSSVTLPHQVVDNP with a Cys added to the N-terminus to allow coupling to KLH. |
Conjugate | FITC |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AC-III; AC3; AC3EC 4.6.1.1 AC-III adenylate cyclase 3 adenylate cyclase type 3 Adenylate cyclase type III Adenylyl cyclase 3 adenylyl cyclase type III ATP pyrophosphate-lyase 3 EC 4.6.1 KIAA0511 olfactive type; Adenylate cyclase type 3; Adenylate cyclase type III; Adenylate cyclase, olfactive type; Adenylyl cyclase 3; adenylyl cyclase, type III; ATP pyrophosphate-lyase 3
Gene Aliases: AC-III; AC3; ADCY3; KIAA0511
UniProt ID: (Human) O60266
Entrez Gene ID: (Human) 109
Molecular Function: adenylate cyclase cyclase guanylate cyclase lyase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.