Novus Biologicals
Manufacturer Code:NBP292583100UL
Catalog # NB027036
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 470-610 of human ADCY1 (NP_066939.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 3&prime-5&prime-cyclic AMP synthetase AC1 adenyl cyclase adenylate cyclase 1 (brain) adenylate cyclase type 1 Adenylate cyclase type I Adenylyl cyclase 1 ATP pyrophosphate-lyase 1 Ca(2+)/calmodulin-activated adenylyl cyclase EC 4.6.1.1; 3',5'-cyclic AMP synthetase; adenyl cyclase; adenylate cyclase 1 (brain); Adenylate cyclase type 1; Adenylate cyclase type I; Adenylyl cyclase 1; ATP pyrophosphate-lyase 1; Ca(2+)/calmodulin-activated adenylyl cyclase
Gene Aliases: AC1; ADCY1; DFNB44
UniProt ID: (Human) Q08828
Entrez Gene ID: (Human) 107
Molecular Function:
adenylate cyclase
cyclase
guanylate cyclase
lyase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.