Invitrogen
Manufacturer Code:MA537744
Catalog # PIMA537744
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1103CT3.4.3 |
Immunogen | recombinant protein between 1-345 amino acids from human ADK |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 2310026J05Rik 5033405D03Rik Adenosine 5'-phosphotransferase adenosine kinase ADK AI255373 AI987814 Ak testicular tissue protein Li 14; Adenosine 5'-phosphotransferase; Adenosine kinase; AK; testicular tissue protein Li 14
Gene Aliases: ADK; AK
UniProt ID: (Human) P55263
Entrez Gene ID: (Human) 132
Molecular Function: carbohydrate kinase kinase nucleotide kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.