Invitrogen
Manufacturer Code:PA575236
Catalog # PIPA575236
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit full length Human Adenosine Kinase. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Adenosine 5'-phosphotransferase; Adenosine kinase; Adenosine kinase Short name&equalsAK Adenosine 5&prime-phosphotransferase ADK; AK; testicular tissue protein Li 14
Gene Aliases: 2310026J05Rik; 5033405D03Rik; ADK; AI255373; AI987814; AK
UniProt ID: (Human) P55263, (Mouse) P55264, (Rat) Q64640
Entrez Gene ID: (Human) 132, (Mouse) 11534, (Rat) 25368
Molecular Function: carbohydrate kinase kinase nucleotide kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.