Novus Biologicals
Manufacturer Code:NBP300364AF750
Catalog # NB065766
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with purified recombinant Adenosine Deaminase/ADA (P00813 Met1-Leu363). |
Conjugate | Alexa Fluor 750 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ADA1 adenine deaminase Adenosine aminohydrolase adenosine deaminase EC 3.5.4.4; Adenosine aminohydrolase; Adenosine deaminase
Gene Aliases: ADA; ADA1
UniProt ID: (Human) P00813
Entrez Gene ID: (Human) 100
Molecular Function:
deaminase
hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.