Novus Biologicals
Manufacturer Code:NBP215855
Catalog # NBP215855
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein encompassing a sequence within the center region of human CECR1. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ADA2 adenosine deaminase CECR1 ADGFadenosine deaminase 2 cat eye syndrome chromosome region candidate 1 Cat eye syndrome critical region protein 1 CECR2 EC 3.5.4.4 IDGFL; Adenosine deaminase 2; Cat eye syndrome critical region protein 1
Gene Aliases: ADA2; ADGF; CECR1; IDGFL; PAN; SNEDS
UniProt ID: (Human) Q9NZK5
Entrez Gene ID: (Human) 51816
Molecular Function:
deaminase
hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.