Novus Biologicals
Manufacturer Code:NBP139474PEATT594
Catalog # N139474P594
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 7F6-G5-A2 |
Immunogen | Full length recombinant human Adenosine A2a R. [UniProt No. P29274] |
Conjugate | PE/ATTO 594 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. Do not freeze. |
Mouse Monoclonal Antibody
Protein Aliases: adenosine A2 receptor adenosine A2a receptor adenosine receptor A2a adenosine receptor subtype A2a ADORA 2 ADORA2 hA2aR RDC8; Adenosine receptor A2a; adenosine receptor subtype A2a
Gene Aliases: A2aR; ADORA2; ADORA2A; RDC8
UniProt ID: (Human) P29274
Entrez Gene ID: (Human) 135
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.