R&D Systems
Manufacturer Code:MAB9497SP
Catalog # MAB9497SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 599717 |
Immunogen | NS0 mouse myeloma cell line transfected with human Adenosine A2a R Met1-Ser412 Accession # P29274 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse MonoclonalAntibody
For Research Use Only
Protein Aliases: A2aR adenosine A2 receptor Adenosine A2a R adenosine A2a receptor adenosine receptor A2a adenosine receptor subtype A2a ADORA 2 ADORA2 ADORA2A hA2aR RDC8; Adenosine receptor A2a; adenosine receptor subtype A2a
Gene Aliases: A2aR; ADORA2; ADORA2A; RDC8
UniProt ID: (Human) P29274
Entrez Gene ID: (Human) 135
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.