Proteintech
Manufacturer Code:203321AP150UL
Catalog # 50-173-2832
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Adenosine A1 Receptor Fusion Protein Ag14092 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: adenosine A1 receptor Adenosine receptor A1 ADORA1 RDC7; adenosine A1 receptor variant 1; adenosine A1 receptor variant 2; Adenosine receptor A1
Gene Aliases: A1-AR; A1AR; A1R; AA1R; ADORA1; AI848715; BB176431; RDC7; Ri
UniProt ID: (Human) P30542, (Mouse) Q60612
Entrez Gene ID: (Human) 134, (Mouse) 11539
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.