Novus Biologicals
Manufacturer Code:NBP292642100UL
Catalog # NB027153
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 40-298 of human SLC25A4 (NP_001142.2). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AAC1 Adenine nucleotide translocator 1 ADP ADP/ATP translocase 1 ANT 1 ANT1adenine nucleotide translocator 1 (skeletal muscle) ATP carrier protein 1 ATP carrier protein heart/skeletal muscle ATP carrier protein heart/skeletal muscle isoform T1 heart/skeletal muscle ATP/ADP translocator PEO2 PEO3 solute carrier family 25 (mitochondrial carrier adenine nucleotidetranslocator) member 4 Solute carrier family 25 member 4 T1ANT; Adenine nucleotide translocator 1; adenine nucleotide translocator 1 (skeletal muscle); ADP,ATP carrier protein 1; ADP,ATP carrier protein, heart/skeletal muscle; ADP,ATP carrier protein, heart/skeletal muscle isoform T1; ADP/ATP translocase 1; ANT 1; heart/skeletal muscle ATP/ADP translocator; solute carrier family 25 (mitochondrial carrier; adenine nucleotide translocator), member 4; Solute carrier family 25 member 4
Gene Aliases: AAC1; ANT; ANT 1; ANT1; MTDPS12; PEO2; PEO3; PEOA2; SLC25A4; T1
UniProt ID: (Human) P12235
Entrez Gene ID: (Human) 291
Molecular Function:
RNA binding protein
amino acid transporter
calcium-binding protein
calmodulin
intracellular calcium-sensing protein
mitochondrial carrier protein
nucleic acid binding
ribosomal protein
transfer/carrier protein
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.