Abnova
Manufacturer Code:PAB10359
Catalog # 89-114-804
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Rat |
Host / Isotype | Sheep |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant GST fusion protein corresponding to amino acids 620-1597 of rat Adarb1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Sheep polyclonal antibody raised against partial recombinant Adarb1.
For Research Use Only
Protein Aliases: Adar2; Double-stranded RNA-specific editase 1; dsRNA adenosine deaminase; Red1; RNA editing deaminase of glutamate receptors; RNA-editing deaminase 1; RNA-editing enzyme 1
Gene Aliases: Adar2; Adarb1; Red1
UniProt ID: (Rat) P51400
Entrez Gene ID: (Rat) 25367
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.