Abnova
Manufacturer Code:MAB18345
Catalog # 89-338-270
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | S46-30 |
Immunogen | Recombinant protein corresponding to amino acids 757-857 at C-terminus of mouse Adam22. |
Conjugate | R-PE |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 2900022I03Rik; a disintegrin and metalloprotease domain (ADAM) 22; ADAM 22; AI854032; Disintegrin and metalloproteinase domain-containing protein 22; MDC2
Gene Aliases: 2900022I03Rik; Adam22; AI854032; MDC2
UniProt ID: (Mouse) Q5TLK3
Entrez Gene ID: (Mouse) 11496
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.