R&D Systems
Manufacturer Code:AF637
Catalog # AF637
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
CyTOF (CyTOF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | <EM>S. frugiperda</EM> insect ovarian cell line <EM>Sf</EM> 21-derived recombinant human Activin RIA/ALK-2 Asp23-Glu123 Accession # Q04771 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: activin A receptor type I; activin A receptor type I ActivinRIA ACTRI ACVR1 ACVR1A ACVRLK2 ALK2 ALK-2 FOP SKR1 TSRI; activin A receptor, type I; activin A receptor, type II-like kinase 2; Activin receptor type I; Activin receptor type-1; Activin receptor-like kinase 2; ACTR-I; ALK-2; hydroxyalkyl-protein kinase; Serine/threonine-protein kinase receptor R1; SKR1; TGF-B superfamily receptor type I; TSR-I
Gene Aliases: ACTRI; ACVR1; ACVR1A; ACVRLK2; ALK2; FOP; SKR1; TSRI
UniProt ID: (Human) Q04771
Entrez Gene ID: (Human) 90
Molecular Function:
TGF-beta receptor
cytokine receptor
kinase
protein kinase
protein kinase receptor
receptor
serine/threonine protein kinase receptor
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.