Novus Biologicals
Manufacturer Code:NBP212010R
Catalog # NBP212010R
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | MM0075-9M29 |
Immunogen | Human Inhibin beta B |
Conjugate | DyLight 550 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: activin AB beta polypeptide; activin AB beta polypeptide Activin beta-B chain INHBB inhibin beta B chain inhibin beta B inhibin beta B (activin AB beta polypeptide) Inhibin beta-2; Activin beta-B chain; Inhibin beta B chain; inhibin, beta B; Inhibin, beta-2
Gene Aliases: INHBB
UniProt ID: (Human) P09529
Entrez Gene ID: (Human) 3625
Molecular Function: growth factor signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.