Invitrogen
Manufacturer Code:PA544198
Catalog # PIPA544198
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human INHBA |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: activin A; Activin beta-A chain; EDF; erythroid differentiation factor; Erythroid differentiation protein; follicle-stimulating hormone-releasing protein; FSH-releasing protein; Inhibin beta A chain; Inhibin beta A EDF FRP; inhibin beta A protein; inhibin beta A subunit; inhibin beta-subunit; inhibin, beta A; inhibin, beta A (activin A, activin AB alpha polypeptide); Inhibin, beta-1; unnamed protein product
Gene Aliases: EDF; FRP; INHB; INHBA
UniProt ID: (Human) P08476, (Pig) P03970, (Horse) P55102, (Bovine) P07995, (Sheep) P43032, (Rat) P18331, (Mouse) Q04998
Entrez Gene ID: (Human) 3624, (Pig) 397093, (Rabbit) 100354786, (Horse) 100034076, (Bovine) 281867, (Dog) 483245, (Goat) 100860960, (Sheep) 443524, (Rat) 29200, (Mouse) 16323
Molecular Function: growth factor signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.