Invitrogen
Manufacturer Code:53649682
Catalog # 50-112-4609
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | HHF35 |
Conjugate | Alexa Fluor 488 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C store in dark DO NOT FREEZE! |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: actin actin alpha 1 actin alpha cardiac actin alpha cardiac 1 actin alpha cardiac muscle 1 actin alpha skeletal muscle actin alpha cardiac 1 actin alpha vascular smooth muscle actin aortic smooth muscle alpha actin 1 alpha actin 2 alpha skeletal muscle alpha skeletal muscle actin alpha-actin cardiac alpha-actin-1 alpha-actin-2 alpha-cardiac actin alpha-sma alphac-actin cell growth-inhibiting gene 46 protein nemaline myopathy type 3 skeletal alpha actin smooth muscle alpha-actin; actin alpha 1; actin alpha cardiac; actin alpha cardiac 1; actin, alpha 2, smooth muscle, aorta; Actin, alpha cardiac muscle 1; Actin, alpha cardiac muscle 1, intermediate form; Actin, alpha skeletal muscle; Actin, alpha skeletal muscle, intermediate form; actin, alpha, cardiac 1; actin, alpha, vascular smooth muscle; Actin, aortic smooth muscle; Actin, aortic smooth muscle, intermediate form; alpha actin; alpha actin 1; alpha actin 2; alpha-actin cardiac; Alpha-actin-1; Alpha-actin-2; Alpha-cardiac actin; alpha-smooth muscle actin; alphac-actin; B-actin; beta actin; cardiac actin; cardiac alpha actin; cardiac muscle alpha actin 1; Cell growth-inhibiting gene 46 protein; nemaline myopathy type 3; skeletal alpha actin; skeletal muscle alpha-actin 1; smooth muscle alpha-actin; vascular smooth muscle alpha-actin
Gene Aliases: 0610041G09Rik; a-SMA; AA959943; AAT6; ACT-4; ACTA; Acta-2; ACTA1; ACTA2; ACTC; Actc-1; ACTC1; actin; Acts; ACTSA; Actsk-1; ACTVS; alphaSMA; ASD5; ASMA; CFTD; CFTD1; CFTDM; CMD1R; CMH11; GIG46; LVNC4; MPFD; MYMY5; NEM1; NEM2; NEM3; SHPM; SMalphaA
UniProt ID: (Human) P68133, (Bovine) P68138, (Pig) P68137, (Bovine) P62739, (Human) P62736, (Human) P68032, (Bovine) Q3ZC07, (Mouse) P68134, (Rat) P68136, (Rat) P62738, (Mouse) P62737, (Mouse) P68033, (Rat) P68035
Entrez Gene ID: (Horse) 100065454, (Human) 58, (Bovine) 281592, (Pig) 100154254, (Dog) 488984, (Pig) 733615, (Bovine) 515610, (Horse) 100062175, (Human) 59, (Dog) 477587, (Human) 70, (Dog) 478250, (Bovine) 533219, (Horse) 100057678, (Pig) 100152267, (Guinea pig) 100716933, (Mouse) 11459, (Rat) 29437, (Rat) 81633, (Mouse) 11475, (Guinea pig) 100722917, (Guinea pig) 100731953, (Mouse) 11464, (Rat) 29275
Molecular Function:
actin and actin related protein
actin family cytoskeletal protein
cytoskeletal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.