Invitrogen
Manufacturer Code:PA554513
Catalog # PIPA554513
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human Actin-like 7B |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: actin-like 7-beta; actin-like 7B; actin-like 7B ACTL7B MGC151192 MGC151194 Tact1 actin-like 7-beta t-actin 1; Actin-like protein 7B; Actin-like-7-beta; testis tissue sperm-binding protein Li 43a
Gene Aliases: ACTL7B; Tact1
UniProt ID: (Human) Q9Y614
Entrez Gene ID: (Human) 10880
Molecular Function:
actin and actin related protein
actin family cytoskeletal protein
cytoskeletal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.