Invitrogen
Manufacturer Code:PA5113560
Catalog # PIPA5113560
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Human Actin-like protein 7B protein (1-415AA) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: actin like 7B actin-like 7B actin-like 7-beta Actin-like protein 7B Actin-like-7-beta ACTL7B BOS_9318 MGC151192 MGC151194 RP11-3J11.2 Tact1 t-actin 1 T-actin-1 testis tissue sperm-binding protein Li 43a Testis-specific actin-1; actin-like 7-beta; actin-like 7B; Actin-like protein 7B; Actin-like-7-beta; t-actin 1; T-actin-1; testis tissue sperm-binding protein Li 43a; Testis-specific actin-1
Gene Aliases: ACTL7B; Tact1
UniProt ID: (Human) Q9Y614, (Mouse) Q9QY83
Entrez Gene ID: (Human) 10880, (Mouse) 11471
Molecular Function:
actin and actin related protein
actin family cytoskeletal protein
cytoskeletal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.